Learn More
Sartorius Stedim Biotech™ Cellulose Acetate Membrane Filter
Sartorius™ Cellulose Acetate Membrane Filter combines high flow rates and thermal stability with very low adsorption characteristics.
1224.00 NOK - 1640.00 NOK
Specifications
Material | Cellulose Acetate |
---|---|
For Use With (Application) | Pressure filtration |
Thickness (Metric) | 120 mm |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
15932450
|
Sartorius Stedim Biotech™
11106--37------N |
0.45 μm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15922450
|
Sartorius Stedim Biotech™
11105--90------G |
0.65 μm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Well suited for use in pressure filtration devices.
- Hydrophilic
- High flow rates thanks to their symmetric structure.
- Compatible with aqueous solutions (pH 4-8), oils, alcohols and other organic solvents.
- Available in different pore sizes from 0.2 to 5μm.
Specifications
Cellulose Acetate | |
Pressure filtration | |
120 mm |