Learn More
Thermo Scientific™ CellSensor™ SIE-bla HEK293T Cell Line
Description
The cell line was created through sorting by FACS of cells responsive to stimulation of the SIE pathway with interleukin 6 (IL-6). The cell line was validated for DMSO tolerance, incubation time with stimulant, and substrate loading conditions. The CellSensor™ SIE-bla HEK 293T cell line responds to agonist treatment as expected from literature and can be adapted for high throughput screening for agonists or antagonists of the SIE pathway with compound libraries. Candidate drugs can also be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Quantity | 1 vial |
| Type | Cell Line |
| Cell Line | HEK 293T |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |