Learn More
Corning™ CellCube™ Culture System Clean Room Cart with Tray
Shop All Corning Products
Click to view available options
:
CellCube Culture System Clean Room Cart with Tray; Stainless steel
Description
- Stainless steel
- Autoclavable
- Mobile rack that can hold upto 4 CellCube™ modules in a small footprint
- Modules can be rotated 360 degrees
- Adjustable wheelbase can be sized to fit a wide selection of tracks found in incubation or environmental chambers making it a versatile product
Specifications
Specifications
| Material | Stainless Steel |
| For Use With (Equipment) | CellCube™ modules |