Learn More
STEMCELL Technologies CellAdhere™ Laminin-521
Product Code. 17178001
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
Product Code. Quantity unitSize
17178001 500 μg 500µg
1 options
Product Code. 17178001
Brand: STEMCELL Technologies 2000117
Matrix for maintenance of human ES and iPS cells in combination with TeSR™ maintenance media
  • Support the growth and differentiation of human embryonic stem (ES) and induced pluripotent stem (iPS) cells under feeder-free conditions with this defined and xeno-free cell culture matrix
  • For consistent cell populations and reproducible results in downstream applications, use CellAdhere™ Laminin-521 with TeSR™ maintenance media to provide a defined culture substrate for cell maintenance
  • Laminin 521 is expressed and secreted by human pluripotent stem cells (hPSCs) in the inner cell mass of the embryo and therefore creates a biologically relevant hPSC culture environment in vitro
  • Compared to other matrices, CellAdhere™ Laminin-521 increases single-cell attachment and survival and does not require the addition of apoptotic inhibitors during plating
  • Note that single-cell passaging of human ES and iPS cells can result in selective pressure and lead to genetic aberrations
  • If passaging as single cells, we recommend checking the karyotype frequently
Specifications
Cell Type Pluripotent Stem Cells
Quantity 500 μg