Learn More
Biochrom Micro-Volume Cell Holder
Product Code. 11300392
Change view
Click to view available options
Type:
Cell Holder
Unit Size:
Each
Product Code. Type unitSize
11300392 Cell Holder Each
1 options
Product Code. 11300392
Brand: Biochrom 80210609
Micro-Volume Cell Holder is for use with 50 μL cuvettes. Accessory for the Ultrospec 7500 and 2100 Pro Spectrophotometer.
  • Micro-volume cell holder
  • For the Ultrospec 2100 Pro Spectrophotometer
  • Use with 50 μL cuvette (80-2076-38)
Specifications
Compatibility Ultrospec 2100 Pro, 3100, 3300, 4300, 5300, and 6300 Models
Description Cell Holder
Type Cell Holder
Disposable Reusable
For Use With (Equipment) Ultrospec 7500, 2100 Pro, 3100, 3300, 4300, 5300, and 6300 Models
Shape Round