Learn More
R&DSystems™ Cell Freezing Medium - II
Product Code. 18412050
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18412050 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18412050 Supplier R&DSystems™ Supplier No. B71295
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Cell Freezing Medium-II contains 90% Fetal Bovine Serum and 10% DMSO (dimethyl sulfoxide). This cryopreservation medium is supplied as a ready-to-use solution.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Cell Culture
Product Line R&D Systems
Quantity 50 mL
Purity or Quality Grade 0.2 μm filtered
Content And Storage -5 to -20 °C, protected from light