Learn More
TPP™ Cell Culture Microplates
Manufactured from polystyrene for crystal clear transparency. TPP™ Cell Culture Microplates feature opto-mechanical treatment of growth area for optimal cell adhesion.
1123.60 NOK - 2464.50 NOK
Specifications
Bottom Shape | Flat |
---|---|
Material | Polystyrene |
Surface Treatment | Tissue Culture |
Dimensions (L x W x H) | 128 x 86 x 22 mm |
Height (Metric) | 22 mm |
Product Code | Brand | Well Count | Quantity | Culture Area | Capacity (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Well Count | Quantity | Culture Area | Capacity (Metric) | Price | Quantity & Availability | ||
11391704
|
TPP™
92024 |
24 | 1/Bag | 1.86 sq. cm | 3.18 mL |
2464.50 NOK
Pack of 126 |
In Stock
Log in to see stock. |
||
11371704
|
TPP™
92006 |
6 | 1/Bag | 9.026 cm2 | 15.53 mL |
2199.50 NOK
Pack of 126 |
In Stock
Log in to see stock. |
||
11381704
|
TPP™
92012 |
12 | 1/Bag | 3.46 sq. cm | 5.96 mL |
2199.50 NOK
Pack of 126 |
Estimated Shipment: 30-04-2024
Log in to see stock. |
||
11391764
|
TPP™
92406 |
6 | 162/Cs. | 9.02 sq. cm | 15.53 mL |
1261.40 NOK
Pack of 72 |
In Stock
Log in to see stock. |
||
11301774
|
TPP™
92412 |
12 | 4/Bag | 3.46 sq. cm | 5.96 mL |
1123.60 NOK
Pack of 72 |
In Stock
Log in to see stock. |
||
Description
Manufactured from polystyrene for crystal clear transparency. TPP™ Cell Culture Microplates feature opto-mechanical treatment of growth area for optimal cell adhesion.
- Air-venting system in lid guarantees controlled gas exchange with low evaporation; vents in plate base prevent condensation and provide consistent air-flow between stacked plates
- Sloped edge allows placement of the lid in one position only; yellow marking area on lid and plate ensure correct lid orientation
- Ridged grip area ensures a secure grasp and prevents accidental lifting of lid
- Easy to see black alpha-numerical labelling between the wells for easy orientation during microscopy
- Growth area on spherical zone only; absolutely flat growth surface
- Plate sizes (6, 12, 24, 48 and 96-well) stack securely together
- Manufactured meeting the standards ANSI/SLAS 1-2004 through ANSI/SLAS 4-2004
- White base colour is embossed firmly on the test plate eliminating loss of identification through detachment
- 2 well geometries available: F-base and U-base
Specifications
Flat | |
Tissue Culture | |
22 mm |
Polystyrene | |
128 x 86 x 22 mm |