Learn More
Biochrom Spare 8 Position Cell Changer
Product Code. 11360392
Change view
Click to view available options
Type:
8-position Carousel
Unit Size:
Each
Product Code. Type unitSize
11360392 8-position Carousel Each
1 options
Product Code. 11360392
Brand: Biochrom 80210801
Spare 8 Position Cell Changer for Ultrospec 7500, 2100 Pro, 3100, 3300, 4300, 5300 and 6300 Models. 10 mm pathlength.
  • Cell changer for 8 x 10 mm pathlength cells
  • Compatible with Ultrospec 7500, 2100 Pro, 3100, 3300, 4300, 5300 and 6300 Models
Specifications
Compatibility Ultrospec 7500, 2100 Pro, 3100, 3300, 4300, 5300, and 6300 Models
Description 8-Cell Changer carousel
Type 8-position Carousel
Disposable Reusable
For Use With (Equipment) Ultrospec 7500, 2100 Pro, 3100, 3300, 4300, 5300, and 6300 Models
Pathlength 10 mm
Shape Round