Learn More
Celite™, for analysis, high purity analytical grade
Product Code. p-2691184 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass Bottle
Unit Size:
10g
25g
Product Code. Quantity Packaging unitSize
10652023 10 g Glass Bottle 10g
10295890 25 g Glass Bottle 25g
2 options
Product Code. 10652023
Brand: Thermo Scientific Acros 173460100
Request Bulk or Custom Format
CAS: 61790-53-2 | O2Si | 60.083 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 61790-53-2
Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
MDL Number MFCD00011232
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym Infusorial earth, Diatomite, Diatomaceous earth
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O
Specifications
Physical Form Powder
Density <0.25 g/mL (centrifuged wet density)
Color Gray/Pink to White
Quantity 10 g
pH 5 to 8 (25°C, 10% aq. suspension)
Loss on Drying 0.5% max.
Loss on Ignition 2% max.
Packaging Glass Bottle
Permeability 0.15 to 0.30 DARCY
Silicon Dioxide 98.7%
Formula Weight 60.08
Grade Analytical
Chemical Name or Material Celite™
Show More Show Less
compliance-icons compliance-icons
Produktidentifikator
  • Celite
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
Sicherheitshinweis
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Ergänzende Informationen
  • MIXTURE LIST-Contains : Diatomaceous earth