missing translation for 'onlineSavingsMsg'
Learn More

Cefoperazone sodium salt, MP Biomedicals™

Product Code. 11445335 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11445335

Brand: MP Biomedicals 0219969501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cefoperazone sodium salt USP, White powder

  • It is used to study drug-protein binding, expression and inhibition of penicillin-binding proteins (PBPs) during cell wall synthesis.
  • Cefoperazone sodium salt is a broad spectrum third generation cephalosporin antibiotic that has similar antimicrobial activity to ceftazidime.
  • It is active against Pseudomonoas aeruginosa and it has enhanced activity against Enterobacteriaceae and Bacteroides sp. in the presence of the beta-lactamase inhibitor sulbactam. Compared to cefotaxime, cefoperazone has greater susceptibility to hydrolysis by some beta-lactamases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62893-20-3
Molecular Formula C25H26N9NaO8S2
Molecular Weight (g/mol) 667.65
MDL Number MFCD07793331
InChI Key NCFTXMQPRQZFMZ-WERGMSTESA-M
PubChem CID 23663974
ChEBI CHEBI:3494

Specifications

Color White
pH 4.5 to 6.5 (1 in 4)
Quantity 1 g
Beilstein 4902135
Synonym Cefobid, Cefoperazone sodium, CPZ, T-1551
Solubility Information Soluble in water (50mg/mL - clear, faint yellow solution).
Grade USP
Physical Form Powder
Assay 870 - 1015 µg/mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.