Learn More
Melford Cefoperazone
Description
Cefoperazone Sodium Salt is a third-generation cephalosporin antibiotic. Cefoperazone inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs).
Cefoperazone is effective against a broad range of bacteria, both Gram-positive and Gram-negative. Cefoperazone Sodium Salt is often used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion allows for the specific growth of bacteria lacking resistance to cefoperazone.
Specifications
Specifications
| Chemical Name or Material | Cefoperazone |
| CAS | 62893-20-3 |
| Quantity | 10 g |
| Molecular Formula | C25H26N9NaO8S2 |