Learn More
Cefixime Trihydrate, Mikromol™
Description
Cefixime Trihydrate
Specifications
Specifications
| Chemical Name or Material | Cefixime Trihydrate |
| CAS | 125110-14-7 |
| Analyte or Component Names | Cefixime Trihydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H15 N5 O7 S2 . 3 H2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Cefixime Trihydrate, Cefixime, (6R,7R)-7-[[(Z)-2-(2-Aminothiazol-4-yl)-2-[(carboxymethoxy)imino]acetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid trihydrate |
| InChI Formula | InChI=1S/C16H15N5O7S2.3H2O/c1-2-6-4-29-14-10(13(25)21(14)11(6)15(26)27)19-12(24)9(20-28-3-8(22)23)7-5-30-16(17)18-7;;;/h2,5,10,14H,1,3-4H2,(H2,17,18)(H,19,24)(H,22,23)(H,26,27);3*1H2/b20-9-;;;/t10-,14-;;;/m1.../s1 |
| SMILES | O.O.O.Nc1nc(cs1)\C(=N\OCC(=O)O)\C(=O)N[C@H]2[C@H]3SCC(=C(N3C2=O)C(=O)O)C=C |
| IUPAC Name | (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(carboxymethoxyimino)acetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;trihydrate |
| Show More |