Learn More
Cefazolin Sodium Salt, TRC
Product Code. 17972515
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17972515 100 mg 100mg
17953805 250 mg 250mg
17940935 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17972515 Supplier Toronto Research Chemicals Supplier No. TRCC242500100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 27164-46-1
Molecular Formula C14 H13 N8 O4 S3 . Na
Molecular Weight (g/mol) 476.49
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[[(5-methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-7-[(1H-tetrazol-1-ylacetyl)amino]-, monosodium salt, (6R,7R)- (9CI), 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[[(5-methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-7-[(1H-tetrazol-1-ylacetyl)amino]-, monosodium salt, (6R-trans)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[[(5-methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-7-[2-(1H-tetrazol-1-yl)acetamido]-, monosodium salt (8CI), 3-[[(5-Methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-7-[2-(1H-tetrazol-1-yl)-acetamido]-3-cephem-4-carboxylic acid sodium salt, 3-[[(5-Methyl-1,3,4-thiadiazol-2-yl)thio]methyl]-8-oxo-7-[2-(1H-tetrazol-1-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid sodium salt, Acef, Ancef, Atirin, Azolin, Biazolina, Bor-Cefazol, Cefacidal, Cefamedin, Cefazil, Cefazina, Cefazoliin, Cefazolin sodium, Cefazolin sodium salt, Cefazoline sodium, Cefazorin DC, Cephazolin sodium, Cephazolin sodium salt, Firmacef, Gramaxin, Iespor, Kefzol, Lampocef, Liviclina, Monosodium cafazolin, NSC 291561, Reflin, SKF 41558, Sodium CEZ, Sodium cefazolin, Sodium cephazolin, Totacef, Zolicef
IUPAC Name sodium;(6R,7R)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES [Na+].Cc1nnc(SCC2=C(N3[C@H](SC2)[C@H](NC(=O)Cn4cnnn4)C3=O)C(=O)[O-])s1
Spezifikation
Quantity 100 mg
InChI Formula InChI=1S/C14H14N8O4S3.Na/c1-6-17-18-14(29-6)28-4-7-3-27-12-9(11(24)22(12)10(7)13(25)26)16-8(23)2-21-5-15-19-20-21;/h5,9,12H,2-4H2,1H3,(H,16,23)(H,25,26);/q;+1/p-1/t9-,12-;/m1./s1
Formula Weight 476.012
Percent Purity >95%
Chemical Name or Material Cefazolin Sodium
Grade HPLC