missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CEACAM6 (Human) Recombinant Protein
Human CEACAM6 full-length ORF ( AAH05008.1, 1 a.a. - 344 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004680-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MGPPSAPPCRLHVPWKEVLLTASLLTFWNPPTTAKLTIESTPFNVAEGKEVLLLAHNLPQNRIGYSWYKGERVDGNSLIVGYVIGTQQATPGPAYSGRETIYPNASLLIQNVTQNDTGFYTLQVIKSDLVNEEATGQFHVYPELPKPSISSNNSNPVEDKDAVAFTCEPEVQNTTYLWWVNGQSLPVSPRLQLSNGNMTLTLLSVKRNDAGSYECEIQNPASANRSDPVTLNVLYGPDVPTISPSKANYRPGENLNLSCHAASNPPAQYSWFINGTFQQSTQELFIPNITVNNSGSYMCQAHNSATGLNRTTVTMITVSGSAPVLSAVATVGITIGVLARVALI
Specifications
AAH05008.1 | |
carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific cross reacting antigen) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CEACAM6 | |
GST |
4680 | |
Wheat germ expression system | |
10 μg | |
CD66c, CEAL, NCA | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |