missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM16 Antibody (2747B) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB9885JF669
This item is not returnable.
View return policy
Description
CEACAM16 Monoclonal antibody specifically detects CEACAM16 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry
Specifications
| CEACAM16 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| carcinoembryonic antigen-related cell adhesion molecule 16 | |
| Chinese Hamster Ovary cell line CHO-derived human CEACAM-16, Met1-Gly425, Accession # Q2WEN9 | |
| RUO | |
| 388551 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, Immunocytochemistry | |
| 2747B | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction