Learn More
Vileda Professional™ CE Tank Cleaner
Product Code. 17862466
Change view
Click to view available options
Autoclavable:
Autoclavable
Unit Size:
Each
Product Code. Autoclavable unitSize
17862466 Autoclavable Each
1 options
Product Code. 17862466
Brand: Vileda Professional™ 162234
Only null left
Add to basket
Add to basket
Specialty above-the-floor tools are ideal for surfaces that are difficult to reach, clean and disinfect. Tools are easy to use and ergonomic allowing full range of motion during cleaning and disinfecting
  • Ideal for inner and outer tank walls
  • Manufactured of electro-polished stainless steel
  • Completely autoclavable
  • Frame is pre-bent to 9 in. radius but is easily adjusted, and can be flat to use on isolators
  • 4 degree of freedom to ensure good contact with the surface being cleaned
  • Easy to use with handle for tight spaces and can be used with telescopic handle for hard to reach areas
Specifications
Product Type Tank Cleaning Tool
Color Silver
Sterility Non-sterile
Autoclavable Autoclavable
Size 9 in.
Quantity 1
For Use With (Application) Inner and Outer Tanks Walls
Material Electropolished Stainless Steel