missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CDR2L (Human) Recombinant Protein
Human CDR2L full-length ORF (ABZ92326.1, 1 a.a. - 459 a.a.) recombinant protein with GST tag at N-terminal.
Brand: Abnova™ H00030850-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MEDFSAEEEESWYDQQDLEQDLHLAAELGKTLLERNKELEGSLQQMYSTNEEQVQEIEYLTKQLDTLRHVNEQHAKVYEQLDLTARDLELTNHRLVLESKAAQQKIHGLTETIERLQAQVEELQAQVEQLRGLEQLRVLREKRERRRTIHTFPCLKELCTSPRCKDAFRLHSSSLELGPRPLEQENERLQTLVGALRSQVSQERQRKERAEREYTAVLQEYSELERQLCEMEACRLRVQELEAELLELQQMKQAKTYLLGPDDHLAEALLAPLTQAPEADDPQPGRGDDLGAQDGVSSPAASPGHVVRKSCSDTALNAIVAKDPASRHAGNLTLHANSVRKRGMSILREVDEQYHALLEKYEELLSKCRQHGAGVRHAGVQTSRPISRDSSWRDLRGGEEGQGEVKAGEKSLSQHVEAVDKRLEQSQPEYKALFKEIFSRIQKTKADINATKVKTHSSK
Specifications
ABZ92326.1 | |
cerebellar degeneration-related protein 2-like | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDR2L | |
GST |
30850 | |
Wheat germ expression system | |
10 μg | |
HUMPPA | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |