Learn More
CDK7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27027-1-AP-20UL
This item is not returnable.
View return policy
Description
The CDK7 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CDK7. This antibody recognizes human, mouse antigen. The CDK7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| CDK7 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 1022 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |