Learn More
CDK7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27027-1-AP-20UL
 View more versions of this product
Product Code. 16845335
  • 1710.33 NOK / 20µL
Description
The CDK7 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CDK7. This antibody recognizes human, mouse antigen. The CDK7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
CDK7
Polyclonal
Unconjugated
20 μL
1022
IgG
Western Blot, Immunohistochemistry, ELISA
333 μg/mL
Rabbit
Primary
Human, Mouse