Learn More
CDK-IN-2, MedChemExpress
Product Code. 16495498
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
Product Code. 16495498
Brand: Medchem Express HY13033
MedChemExpress CDK-IN-2 is a potent and specific CDK9 inhibitor with IC50 of <8 nM, extracted from reference 1, example 4.
CDK-IN-2 is a potent and specific CDK9 inhibitor with IC50 of <8 nM, extracted from reference 1, example 4.
Specifications
Chemical Name or Material CDK-IN-2
CAS 1269815-17-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H19ClFN3O2
Quantity 10 mg
Synonym CDK inhibitor II
Solubility Information DMSO : ≥ 100 mg/mL (274.87 mM)
SMILES FC1=CC(C2=CC(NC([C@H]3CNCCC3)=O)=NC=C2Cl)=C(C=C1)OC
Molecular Weight (g/mol) 363.81
Formula Weight 363.81
Percent Purity 98.82%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.