Learn More
CDDO-EA, MedChemExpress
Product Code. 16495018
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16495018
Brand: Medchem Express HY12213
MedChemExpress CDDO-EA is an NF-E2 related factor 2/antioxidant response element (Nrf2/ARE) activator.
CDDO-EA is an NF-E2 related factor 2/antioxidant response element (Nrf2/ARE) activator.
Specifications
Chemical Name or Material CDDO-EA
Physical Form Solid
CAS 932730-51-3
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C33H46N2O3
For Use With (Application) Cancer-programmed cell death
Synonym CDDO ethyl amide TP319 RTA 405
Solubility Information DMSO : ≥ 34 mg/mL (65.54 mM)
SMILES O=C1C(C#N)=C[C@@]2(C)[C@](CC[C@]([C@@]3(C)[C@@]4([H])[C@@]5([H])[C@@](CCC(C)(C)C5)(C(NCC)=O)CC3)(C)C2=CC4=O)([H])C1(C)C
Molecular Weight (g/mol) 518.73
Formula Weight 518.73
Percent Purity 98.15%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.