Learn More
CDDO-EA, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
CDDO-EA is an NF-E2 related factor 2/antioxidant response element (Nrf2/ARE) activator.
Specifications
Specifications
| Chemical Name or Material | CDDO-EA |
| Physical Form | Solid |
| CAS | 932730-51-3 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C33H46N2O3 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | CDDO ethyl amide TP319 RTA 405 |
| Solubility Information | DMSO : ≥ 34 mg/mL (65.54 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.