missing translation for 'onlineSavingsMsg'
Learn More

CDDO-EA, MedChemExpress

Product Code. 16457747
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16457747 5 mg 5mg
16495018 10 mg 10mg
16481257 50 mg 50mg
16415776 100 mg 100mg
4 options
This item is not returnable. View return policy

Product Code. 16457747

Brand: Medchem Express HY12213

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CDDO-EA is an NF-E2 related factor 2/antioxidant response element (Nrf2/ARE) activator.

CDDO-EA is an NF-E2 related factor 2/antioxidant response element (Nrf2/ARE) activator.

Specifications

Chemical Name or Material CDDO-EA
Physical Form Solid
CAS 932730-51-3
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C33H46N2O3
For Use With (Application) Cancer-programmed cell death
Synonym CDDO ethyl amide TP319 RTA 405
Solubility Information DMSO : ≥ 34 mg/mL (65.54 mM)
SMILES O=C1C(C#N)=C[C@@]2(C)[C@](CC[C@]([C@@]3(C)[C@@]4([H])[C@@]5([H])[C@@](CCC(C)(C)C5)(C(NCC)=O)CC3)(C)C2=CC4=O)([H])C1(C)C
Molecular Weight (g/mol) 518.73
Formula Weight 518.73
Percent Purity 98.15%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.