missing translation for 'onlineSavingsMsg'
Learn More

CDC25B Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Product Code. 16806005 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16806005 20 μL 20µL
16895995 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16806005

Brand: Proteintech 281091AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The CDC25B antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CDC25B. This antibody recognizes human antigen. The CDC25B antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Antigen CDC25B
Applications Western Blot, Immunohistochemistry, ELISA
Classification Polyclonal
Concentration 533 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 994
Target Species Human
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.