Learn More
CDC25B Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28109-1-AP-150UL
This item is not returnable.
View return policy
Description
The CDC25B antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CDC25B. This antibody recognizes human antigen. The CDC25B antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| CDC25B | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 994 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 533 μg/mL | |
| Rabbit | |
| Primary | |
| Human |