Learn More
CDC25B Mouse anti-Human, Unconjugated, Clone: 1H3A4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67145-1-IG-20UL
 View more versions of this product
Product Code. 16809615
  • 1710.33 NOK / 20µL
Description
The CDC25B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDC25B. This antibody recognizes human, mouse, rat antigen. The CDC25B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
CDC25B
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
1H3A4
Unconjugated
20 μL
994
IgG2b