Learn More
CDC25B Mouse anti-Human, Unconjugated, Clone: 1H3A4, Proteintech
Product Code. 16899605 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16899605 150 μL 150µL
16809615 20 μL 20µL
2 options
Product Code. 16899605
Brand: Proteintech 671451IG150UL
Mouse Monoclonal Antibody
The CDC25B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDC25B. This antibody recognizes human, mouse, rat antigen. The CDC25B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Antigen CDC25B
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1H3A4
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 994
Target Species Human, Rat, Mouse
Isotype IgG2b