missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CDC2 (Human) Recombinant Protein
Human CDC2 full-length ORF ( AAH14563, 1 a.a. - 297 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000983-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MEDYTKIEKIGEGTYGVVYKGRHKTTGQVVAMKKIRLESEEEGVPSTAIREISLLKELRHPNIVSLQDVLMQDSRLYLIFEFLSMDLKKYLDSIPPGQYMDSSLVKSYLYQILQGIVFCHSRRVLHRDLKPQNLLIDDKGTIKLADFGLARAFGIPIRVYTHEVVTLWYRSPEVLLGSARYSTPVDIWSIGTIFAELATKKPLFHGDSEIDQLFRIFRALGTPNNEVWPEVESLQDYKNTFPKWKPGSLASHVKNLDENGLDLLSKMLIYDPAKRISGKMALNHPYFNDLDNQIKKM
Specifications
AAH14563 | |
cell division cycle 2, G1 to S and G2 to M | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDC2 | |
GST |
983 | |
Wheat germ expression system | |
10 μg | |
CDC28A, CDK1, DKFZp686L20222, MGC111195 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |