missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC14B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25187-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
CDC14B is a member of the dual specificity protein tyrosine phosphatase family. This protein is highly similar to Saccharomyces cerevisiae Cdc14, a protein tyrosine phosphatase involved in the exit of cell mitosis and initiation of DNA replication, which suggests the role in cell cycle control. This protein has been shown to interact with and dephosphorylates tumor suppressor protein p53, and is thought to regulate the function of p53. Alternative splice of this gene results in 3 transcript variants encoding distinct isoforms.Specifications
CDC14B | |
Polyclonal | |
Unconjugated | |
CDC14B | |
CDC14B, Cdc14B1, Cdc14B2, CDC14B3, hCDC14B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8555 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60729 | |
CDC14B | |
CDC14B Fusion Protein Ag18227 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |