missing translation for 'onlineSavingsMsg'
Learn More

CDC123 Mouse anti-Human, Clone: OTI1E4, lyophilized, TrueMAB™

Product Code. 17694791
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. Quantity unitSize
17694791 100 μg 100µg
1 options
This item is not returnable. View return policy

Product Code. 17694791

Brand: Origene Technologies CF505649

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Required for S phase entry of the cell cycle.

Specifications

Antigen CDC123
Applications Western Blot
Classification Monoclonal
Clone OTI1E4
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene CDC123
Gene Accession No. O75794
Gene Alias C10orf7, D123
Gene Symbols CDC123
Host Species Mouse
Immunogen Full length human recombit protein of human CDC123 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8872
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.