Learn More
CD90 (Thy-1) Mouse, anti-Human, APC, Clone: 5E10, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60045AZ
Additional Details : Weight : 0.00110kg
 View more versions of this product
Product Code. 17108761
  • 6404.14 NOK / 100 tests
Estimated Shipment: 16-10-2023
Description
  • The 5E10 antibody reacts with CD90 (Thy-1), a GPI-linked membrane glycoprotein that is N-glycosylated at two sites, giving rise to 25 - 37 kDa molecules
  • CD90 has roles in signal transduction, cell adhesion and migration, neurite outgrowth, T cell activation, tumor suppression, and inhibition of the proliferation and differentiation of hematopoietic stem cells
  • It is a known ligand of β2 and β3 integrins and upregulates synthesis of fibronectin, osteonectin and thrombospondin
  • CD90 is broadly expressed, being found on human thymocytes, neurons, some glial cells, fibroblasts, activated endothelial cells, some leukemia cell lines and a distinct subset (<1%) of CD3+CD4+ T cells in human peripheral blood
  • This antibody clone has been verified for labeling human mesenchymal cells grown in MesenCult™ Proliferation Kit (Human; Catalog #05411) and MesenCult™-XF Medium (Catalog #05420)
Specifications
Specifications
CD90 (Thy-1)
Monoclonal
APC
100 Tests
Baboon, Cynomolgus, Rhesus Monkey, Pig, Human
Hematopoietic Stem and Progenitor Cells, Mammary Cells, Mesenchymal Stem and Progenitor Cells
Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry
5E10
Mouse
Primary
IgG1 κ