missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD89 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13245-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the immunoglobulin gene superfamily and encodes a receptor for the Fc region of IgA. The receptor is a transmembrane glycoprotein present on the surface of myeloid lineage cells such as neutrophils, monocytes, macrophages, and eosinophils, where it mediates immunologic responses to pathogens. It interacts with IgA-opsonized targets and triggers several immunologic defense processes, including phagocytosis, antibody-dependent cell-mediated cytotoxicity, and stimulation of the release of inflammatory mediators. Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
CD89 | |
Polyclonal | |
Unconjugated | |
FCAR | |
CD89, FCAR, IgA Fc receptor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2204 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P24071 | |
FCAR | |
CD89 Fusion Protein Ag3928 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |