missing translation for 'onlineSavingsMsg'
Learn More

Origene Technologies CD86 Monoclonal Antibody (OTI8C9), TrueMAB™, OriGene

Mouse Monoclonal Antibody

Brand:  Origene Technologies TA813131

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 17557844

  • 5484.49 NOK / 100µL
Estimated Shipment: 19-06-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD86 Monoclonal antibody specifically detects CD86 in Human samples. It is validated for Flow Cytometry, Western Blot
Specifications

Specifications

CD86
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P42081
Mouse
Protein A/G
RUO
942
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Western Blot
OTI8C9
Unconjugated
CD86
activation B7-2 antigen; B lymphocyte activation antigen B72; B7; B7.2; B70; B7-2; B-lymphocyte activation antigen B7-2; BU63; CD28 antigen ligand 2; Cd28l2; CD28LG2; CD86; CD86 antigen; CD86 antigen (CD28 antigen ligand 2, B7-2 antigen); CD86 antigen precursor; CD86 molecule; CLS1; CTLA-4 counter-receptor B7.2; early T cell costimulatory molecule-1; early T-cell costimulatory molecule 1; Early T-cell co-stimulatory molecule 1; ETC-1; FUN-1; LAB72; Ly58; Ly-58; MB7; MB7-2; membrane glycoprotein; MGC34413; T-lymphocyte activation antigen CD86; TS/A-2
Full length human recombinant protein of human CD86 (NP_787058) produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers