Learn More
CD86 Monoclonal Antibody (OTI8A7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA813085
Encompass_Preferred
 View more versions of this product
Product Code. 17567774
  • 6331.68 NOK / 100µL
Description
CD86 Monoclonal antibody specifically detects CD86 in Human samples. It is validated for Flow Cytometry, Western Blot
Specifications
Specifications
CD86
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P42081
Mouse
Protein A/G
RUO
942
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Western Blot
OTI8A7
Unconjugated
CD86
activation B7-2 antigen; B lymphocyte activation antigen B72; B7; B7.2; B70; B7-2; B-lymphocyte activation antigen B7-2; BU63; CD28 antigen ligand 2; Cd28l2; CD28LG2; CD86; CD86 antigen; CD86 antigen (CD28 antigen ligand 2, B7-2 antigen); CD86 antigen precursor; CD86 molecule; CLS1; CTLA-4 counter-receptor B7.2; early T cell costimulatory molecule-1; early T-cell costimulatory molecule 1; Early T-cell co-stimulatory molecule 1; ETC-1; FUN-1; LAB72; Ly58; Ly-58; MB7; MB7-2; membrane glycoprotein; MGC34413; T-lymphocyte activation antigen CD86; TS/A-2
Full length human recombinant protein of human CD86 (NP_787058) produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.