missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD86 Monoclonal Antibody (OTI8A7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813085S
Additional Details : Weight : 0.05000kg
Description
CD86 Monoclonal antibody specifically detects CD86 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
CD86 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P42081 | |
Mouse | |
Protein A/G | |
RUO | |
942 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
OTI8A7 | |
Unconjugated | |
CD86 | |
activation B7-2 antigen; B lymphocyte activation antigen B72; B7; B7.2; B70; B7-2; B-lymphocyte activation antigen B7-2; BU63; CD28 antigen ligand 2; Cd28l2; CD28LG2; CD86; CD86 antigen; CD86 antigen (CD28 antigen ligand 2, B7-2 antigen); CD86 antigen precursor; CD86 molecule; CLS1; CTLA-4 counter-receptor B7.2; early T cell costimulatory molecule-1; early T-cell costimulatory molecule 1; Early T-cell co-stimulatory molecule 1; ETC-1; FUN-1; LAB72; Ly58; Ly-58; MB7; MB7-2; membrane glycoprotein; MGC34413; T-lymphocyte activation antigen CD86; TS/A-2 | |
Full length human recombinant protein of human CD86 (NP_787058) produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |