Learn More
CD84 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564444
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RRQGRIFPED AASKKTIYTY IMASRNTQPA ESRIYDEILQ SKVLPSKEEP VNTVYSEVQF ADKMGKASTQ DSKPPGTS Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 53%.
CD84 is expressed on mature B cells and on B cell lines, including pre B cell lines, but not on plasma cell lines. Immunohistochemical studies demonstrated that CD84 strongly expressed on tissues macrophages. CD84 is also highly expressed on platelets and, at low levels on peripheral blood T cells. It is a highly glycosylated protein and belongs to immunoglobulin superfamily. It may play a role in leukocyte activation.Specifications
CD84 | |
Polyclonal | |
Unconjugated | |
CD84 | |
A130013D22Rik; CD84; CD84 antigen; CD84 antigen (leukocyte antigen); CD84 molecule; CDw84; cell surface antigen MAX.3; DKFZp781E2378; hCD84; Hly9-beta; leucocyte differentiation antigen CD84; leukocyte antigen CD84; leukocyte differentiation antigen CD84; LY9B; mCD84; RGD1564553; Signaling lymphocytic activation molecule 5; SLAM family member 5; SLAMF5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8832 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UIB8 | |
CD84 | |
Recombinant protein corresponding to Human CD84 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only