Description
The CD81 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD81. This antibody recognizes human antigen. The CD81 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
Specifications
| Antigen | CD81 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | 1G2C6 |
| Concentration | 1000 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 975 |
| Show More |