missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CD80 (Human) Recombinant Protein

Product Code. 16122841
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16122841 10 μg 10µg
16132841 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16122841 Supplier Abnova™ Supplier No. H00000941P01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human CD80 full-length ORF ( AAH42665, 35 a.a. - 288 a.a.) recombinant protein with GST-tag at N-terminal.

  • Sequence: VIHVTKEVKEVATLSCGHNVSVEELAQTRIYWQKEKKMVLTMMSGDMNIWPEYKNRTIFDITNNLSIVILALRPSDEGTYECVVLKYEKDAFKREHLAEVTLSVKADFPTPSISDFEIPTSNIRRIICSTSGGFPEPHLSWLENGEELNAINTTVSQDPETELYAVSSKLDFNMTTNHSFMCLIKYGHLRVNQTFNWNTTKQEHFPDNLLPSWAITLISVNGIFVICCLTYCFAPRCRERRRNERLRRESVRPV

Specifications

Accession Number AAH42665
Gene ID (Entrez) 941
Name CD80 molecule
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias CD28LG, CD28LG1, LAB7
Gene Symbol CD80
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.