Learn More
CD80 (B7-1) Monoclonal Antibody (16-10A1), Super Bright™ 645, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Affymetrix eBioscience 64-0801-82
Additional Details : Weight : 0.01000kg
Description
Description: The GL1 monoclonal antibody reacts with mouse CD86, an ∽80 kDa surface receptor also known as B7-2. CD86 and CD80 are members of the B7 family of costimulatory molecules. CD86 is expressed at low level on B cells, macrophages, and dendritic cells and is upregulated on B cells through a variety of surface stimuli including the BCR complex, CD40 and some cytokine receptors. CD86 is also expressed by activated mouse T cells and thioglycolate-elicited peritoneal cells. In addition to CD80 (B7-1), CD86 is a counter-receptor for the T cell surface molecules CD28 and CD152 (CTLA-4). This interaction plays a critical role in T-B crosstalk, T cell costimulation, autoantibody production and Th2-mediated Ig production. The kinetics of upregulation of CD86 upon stimulation, supports its major contribution during the primary phase of an immune response. Applications Reported: This GL1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This GL1 antibody has been tested by flow cytometric analysis . This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
CD80 (B7-1) and CD86 (B7-2) are ligands of T cell critical costimulatory molecule CD28, and of an inhibitory receptor CTLA-4 (CD152). Both B7 molecules are expressed on professional antigen-presenting cells and are essential for T cell activation, and both molecules can also substitute for each other in this process. CD80 is rapidly induced on the surface of in vitro activated B cells, with Epstein Barr Virus (EBV) transformed B cell lines, with Burkitts lymphoma cell lines, with freshly isolated follicular B lymphoma cells, T cells, and monocytes. The B-lymphocyte activation antigen B7-1 provides regulatory signals for T lymphocytes as a consequence of binding to the CD28 and CTLA4 ligands of T cells. Diseases associated with CD80 dysfunction include gallbladder squamous cell carcinoma and myocarditis.Specifications
CD80 (B7-1) | |
Monoclonal | |
0.2 mg/mL | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Q00609 | |
CD80 | |
Affinity chromatography | |
RUO | |
12519, 397161, 403765 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
16-10A1 | |
Super Bright 645 | |
CD80 | |
Activation B7-1 antigen; B7; B7 protein; B7.1; B71; B7-1; B7-1 protein; B7-1 protein precursor; BB1; B-lymphocyte activation antigen B7; Cd28l; CD28LG; CD28LG1; CD80; Cd80 a rat homolog of the human CD28/CTLA - 4 ligand (B7-1); CD80 anitgen; CD80 antigen; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); Cd80 molecule; CD80 protein precursor; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; LAB7; Ly53; Ly-53; MIC17; sCD 80; sCD80; secreted B7-1 protein precursor; soluble CD 80; soluble CD80; T-cell costimulatory molecule; T-cell co-stimulatory protein B7-1; T-lymphocyte activation antigen CD80; TS/A-1; TSA1 | |
Armenian Hamster | |
100 μg | |
Primary | |
Canine, Mouse, Porcine | |
Antibody | |
IgG |
For Research Use Only