missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Mouse anti-Human , Alexa Fluor 647, Clone: HIT8a, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566854
This item is not returnable.
View return policy
Description
CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Flow Cytometry
Specifications
| CD8 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| CD8α; CD8A; CD8 alpha; Leu2a; MAL; T8; p32 | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 647 under optimum conditions, and unreacted Alexa Fluor 647 was removed. |
| Flow Cytometry | |
| HIT8a | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| RUO | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction