missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8/CD38 Mouse anti-Human, FITC,PE, Clone: SK1,HB7, BD Simultest™
Mouse Monoclonal Antibody
Brand: BD Biosciences 349527
This item is not returnable.
View return policy
Description
CD8/CD38 Monoclonal antibody specifically detects CD8/CD38 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD8/CD38 | |
| Monoclonal | |
| FITC/PE | |
| 50 Tests | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |
| Flow Cytometry | |
| SK1, HB7 | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction