missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79A Antibody (rIGA/6986) [Janelia Fluor™ 669], Novus Biologicals™
Description
CD79A Monoclonal specifically detects CD79A in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Specifications
Specifications
| Antigen | CD79A |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | rIGA/6986 |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD79a antigen, CD79A antigen (immunoglobulin-associated alpha), CD79a molecule, immunoglobulin-associated alpha, IGAB-cell antigen receptor complex-associated protein alpha chain, Ig-alpha, MB1, MB-1, MB-1 membrane glycoprotein, Membrane-bound immunoglobulin-associated protein, Surface IgM-associated protein |
| Host Species | Mouse |
| Immunogen | Recombinant fragment (around aa1-100) of human CD79A protein (exact sequence is proprietary) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?