missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD74 Mouse anti-Human, Clone: 2D8A1, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | CD74 |
|---|---|
| Clone | 2D8A1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16858225
|
Proteintech
66390-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16848225
|
Proteintech
66390-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Expression of the invariant chain of the HLA-DR antigen (CD74) by B-lymphocytes begins in the pre-B stage but lost before the final differentiation to plasma cells. This protein is expressed in B-lymphocytes and many B-cell lymphomas.Specifications
| CD74 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CD74, DHLAG, HLADG, Ia GAMMA, Ii, p33 | |
| CD74 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 2D8A1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P04233 | |
| 972 | |
| CD74 Fusion Protein Ag25503 | |
| Primary | |
| -20°C | |
| CD74 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title