Learn More
CD64 Monoclonal Antibody (10.1), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen CD64044
Additional Details : Weight : 0.50000kg
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
CD64 Monoclonal antibody specifically detects CD64 in Human samples. It is validated for Flow Cytometry.Specifications
CD64 | |
Monoclonal | |
PE | |
FCGR1A | |
AI323638; AV092959; CD64; CD64 antigen; CD64A; Fc fragment of IgG high affinity Ib receptor; Fc fragment of IgG receptor Ia; Fc fragment of IgG, high affinity Ia, receptor (CD64); Fc fragment of IgG, high affinity Ia, receptor for (CD64); Fc fragment of IgG, high affinity Ib, receptor for (CD64); Fc gamma receptor; Fc gamma receptor Ia; Fc receptor, IgG, high affinity I; Fcg1; Fc-gamma receptor 1; Fc-gamma receptor I A1; Fc-gamma RI; Fc-gamma RIA; FcgammaRI; FcgammaRIa; Fcgr1; FCGR1A; Fcgr1b; FcgRI; FCRI; FLJ18345; high affinity IgG Fc receptor, subunit beta; high affinity immunoglobulin gamma Fc receptor I; high-affinity immunoglobulin gamma Fc receptor I; IGFR1; IgG Fc receptor I; IgG high affinity Fc receptor; IGGHAFC; RP11-196G18.2 | |
Mouse | |
purified | |
RUO | |
2209 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
10.1 | |
PBS with BSA, sucrose and 0.1% sodium azide | |
P12314 | |
FCGR1A | |
Human CD64 | |
2 mL | |
Primary | |
Human | |
Antibody | |
IgG1 |