missing translation for 'onlineSavingsMsg'
Learn More

CD56 (NCAM) Recombinant Mouse Monoclonal Antibody (CMSSB), Alexa Fluor™ Plus 647, Invitrogen™

Product Code. 30181669
Change view
Click to view available options
Quantity:
20 μL
50 μL
Unit Size:
20µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30181669 20 μL 20µL
30195812 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30181669 Supplier Invitrogen Supplier No. 740024MP64720UG

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Alexa Fluor™ Plus recombinant antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning data. Alexa Fluor™ Plus antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor™ Plus antibodies provide brighter signal compared to leading Alexa Fluor™ antibodies, providing you with better signal-to-noise for your critical experiments. These antibodies show better specificity and lot-to-lot consistency as these are recombinant antibodies, generated by cloning specific genes for the desired antibodies into an expression vector and expressed in vitro. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Applications Tested: This CMSSB antibody has been tested by immunocytochemistry and flow cytometric analysis of NK-92 cells. This may be used for immunocytochemistry at 10 μg/mL and for flow cytometry at less than or equal to 0.25 μg per test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 658 nm; Emission: 675 nm; Laser: Red Laser Filtration: 0.2 μm post-manufacturing filtered.

CD56 (NCAM) Recombinant Monoclonal antibody specifically detects CD56 (NCAM) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD56 (NCAM)
Applications Flow Cytometry, Immunocytochemistry
Classification Recombinant Monoclonal
Clone CMSSB
Concentration 1.0 mg/mL
Conjugate Alexa Fluor Plus 647
Formulation Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.8
Gene Ncam1
Gene Accession No. P13591
Gene Alias adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; Cd56; CD-56; CD56 120 kDa GPI-linked isoform; CD56 140 kDa isoform; CD56 140 kDa VASE isoform; E NCAM; E-NCAM; MSK39; N CAM1; NCAM; N-CAM; Ncam1; N-CAM-1; NCAM-1; NCAMC; NCAM-C; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM
Host Species Mouse
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4684
Target Species Human
Content And Storage 4° C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.