Learn More
CD52 Mouse anti-Human, Unconjugated, Clone: 1B10E8, Proteintech
Product Code. 16878975 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16878975 150 μL 150µL
16888975 20 μL 20µL
2 options
Product Code. 16878975
Brand: Proteintech 667841IG150UL
Mouse Monoclonal Antibody
The CD52 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD52. This antibody recognizes human antigen. The CD52 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Antigen CD52
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1B10E8
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 1043
Target Species Human
Isotype IgG1