Learn More
CD49f (Integrin α6) Rat, anti-Mouse, PE, Clone: GoH3, STEMCELL Technologies
Rat Monoclonal Antibody
Brand:  Stemcell Technologies 60037PE.1
Encompass_Preferred
 View more versions of this product
Product Code. 17138701
  • 3571.72 NOK / 25 tests
Description
  • The GoH3 antibody reacts with CD49f (integrin α6), an ∼150 kDa transmembrane glycoprotein that associates non-covalently with CD29 (integrin β1) or CD104 (integrin β4) to form the heterodimeric receptors VLA-6 and α6β4, which bind the extracellular matrix protein laminin
  • CD49f is a disulfide-linked dimer comprising an ∼120 kDa heavy chain and an ∼30 kDa membrane-bound light chain
  • Splice variants exist, which affect the cytoplasmic domain of the protein
  • CD49f is expressed on the surface of T cells, monocytes, platelets, placental trophoblasts, epithelial cells, and endothelial cells
  • It is involved in cell adhesion and regulating signaling pathways involved in a variety of processes, including the activation and proliferation of T cells, and the differentiation and maintenance of stem cell pluripotency
  • CD49f is considered the most important marker for selecting mouse mammary stem and progenitor cells
  • The GoH3 antibody reacts with an extracellular epitope on CD49f
Specifications
Specifications
CD49f (Integrin α6)
Monoclonal
PE
25 Tests
Baboon, Cynomolgus, Pig, Human
Mammary Cells
Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay
GoH3
Rat
Primary
IgG2a κ