Learn More
CD49d (Integrin alpha 4) Monoclonal Antibody (9F10), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
30070.00 NOK - 150400.00 NOK
Specifications
| Antigen | CD49d (Integrin alpha 4) |
|---|---|
| Clone | 9F10 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Functional Assay |
| Classification | Monoclonal |
Description
The 9F10 monoclonal antibody reacts with human CD49d, the 150 kDa integrin alpha4 subunit. The complex of CD49d non-covalently associated with integrin beta1 (CD29), also known as VLA-4, is a receptor for fibronectin and VCAM-1 (CD106). This complex is expressed by thymocytes, peripheral lymphocytes, monocytes and eosinophils. CD49d also associates with integrin beta7 and binds to the Mucosal Addressin Cell-Adhesion Molecule-1 (MadCAM-1). Applications Reported: The 9F10 antibody has been reported for use in flow cytometric analysis. 9F10 has also been reported in in vitro functional assays. Applications Tested: The 9F10 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/ μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
CD49d (Integrin alpha 4) Monoclonal antibody specifically detects CD49d (Integrin alpha 4) in Human samples. It is validated for Flow Cytometry, Functional AssaySpecifications
| CD49d (Integrin alpha 4) | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| 269C wild type; alpha 4 subunit of VLA-4 receptor; antigen CD49D, alpha-4 subunit of VLA-4 receptor; CD49 antigen-like family member D; CD49d; IA4; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin subunit alpha 4; integrin, alpha 4; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); ITGA4; Itga4B; ITGA-IV; LPAM subunit alpha; lymphocyte Peyer patch adhesion molecules subunit alpha; very late activation protein 4 receptor, alpha 4 subunit; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| 9F10 | |
| Flow Cytometry, Functional Assay | |
| Functional Grade | |
| Mouse | |
| Human | |
| P13612 | |
| 3676 | |
| Primary | |
| 4° C | |
| ITGA4 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.