Learn More
Abnova™ CD48 (Human) Recombinant Protein
Human CD48 full-length ORF ( AAH30224, 1 a.a. - 169 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000962-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MCSRGWDSCLALELLLLPLSLLVTSIQGHLVHMTVVSGSNVTLNISESLPENYKQLTWFYTFDQKIVEWDSRKSKYFESKFKGRVRLDPQSGALYISKVQKEDNSTYIMRVLKKTGNEQEWKIKLQVLGESGEPKSKSPLQWPQMDHCRASWEAWGTLGEEERKTSGQV
Specifications
AAH30224 | |
CD48 molecule | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD48 | |
GST |
962 | |
Wheat germ expression system | |
25 μg | |
BCM1, BLAST, BLAST1, MEM-102, SLAMF2, hCD48, mCD48 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |