missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Origene Technologies CD47 Monoclonal Antibody (OTI6A2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813323
Additional Details : Weight : 0.05000kg
Description
CD47 Monoclonal antibody specifically detects CD47 in Human samples. It is validated for Flow CytometrySpecifications
CD47 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q08722 | |
Mouse | |
Protein A/G | |
RUO | |
961 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry | |
OTI6A2 | |
Unconjugated | |
CD47 | |
9130415E20Rik; AA407862; AI848868; antigen identified by monoclonal antibody 1D8; antigenic surface determinant protein OA3; AW108519; B430305P08Rik; Cd47; CD47 antigen; CD47 antigen (Rh-related antigen, integrin-associated signal transducer); CD47 antigen/integrin-associated protein; CD47 glycoprotein; CD47 molecule; CD47/IAP; IAP; integrin associated protein; integrin-associated protein; integrin-associated signal transducer; Itgp; Leukocyte surface antigen CD47; MER6; OA3; Protein MER6; Rh-related antigen; sCD47; soluble CD 47; soluble CD47 | |
Full length human recombinant protein of human CD47 (NP_001768) produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |