missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD47 Monoclonal Antibody (B6H12), Alexa Fluor™ 488, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
1790.00 NOK - 4505.00 NOK
Specifications
| Antigen | CD47 |
|---|---|
| Clone | B6H12 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17103973
|
Invitrogen
53-0479-41 |
25 Tests | |||||||
17113973
![]() |
Invitrogen™
53-0479-42 |
100 Tests |
4505.00 NOK
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD47 (integrin-associated protein, IAP) is a ubiquitously expressed cell surface transmembrane glycoprotein interacting with several integrins and regulating their functions. Engagement of CD47 by soluble ligands or counter receptors modulates various signaling pathways, such as activation of heterotrimeric G proteins. Functionally, CD47 is involved in the increase in intracellular calcium concentration that occurs upon cell adhesion to extracellular matrix. CD47 is also a receptor for the C-terminal cell binding domain of thrombospondin, and it may play a role in membrane transport and signal transduction. CD47 has broad tissue distribution, is reduced in expression on Rh erythrocytes. CD47 acts as a ligand for CD172a (signal regulatory protein alpha, SIRP alpha), an immune inhibitory receptor on macrophages; this interaction prevents phagocytosis of CD47-positive cells. Moreover, CD47-CD172a system affects cell migration, B cell adhesion, T cell activation, and is involved in modulation of chondrocyte responses to mechanical signals, and promotes neuronal development, being especially abundant in synapse-rich regions of brain and retina.Specifications
| CD47 | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
| 961 | |
| IgG1 κ | |
| Affinity chromatography | |
| CD47 |
| B6H12 | |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| Mouse | |
| Human | |
| Q08722 | |
| CD47 | |
| Primary | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title